Assay Details
Gene Symbol: SPSB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049789.1 | XP_017905278.1 | 2 - 3 | 927 | 63 | |
GenBank mRNA | JO369143.2 | - | 1 - 2 | 250 | 63 | |
JT716596.1 | - | 2 - 3 | 315 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
