Assay Details
| Gene Symbol: TSSK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054628.1 | XP_017910117.1 | 2 - 3 | 1154 | 92 | |
| XM_018054629.1 | XP_017910118.1 | 2 - 3 | 1154 | 92 | ||
| XM_018054630.1 | XP_017910119.1 | 2 - 3 | 640 | 92 | ||
More Information
Set Membership: |
|

TSSK4
Genomic Map