Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063734.1 | XP_017919223.1 | 2 - 3 | 317 | 62 | |
XM_018063735.1 | XP_017919224.1 | 2 - 3 | 313 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
