Assay Details
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062670.1 | XP_017918159.1 | 7 - 8 | 741 | 61 | |
XM_018062675.1 | XP_017918164.1 | 6 - 7 | 644 | 61 | ||
XM_018062681.1 | XP_017918170.1 | 6 - 7 | 630 | 61 | ||
XM_018062688.1 | XP_017918177.1 | 6 - 7 | 670 | 61 | ||
XM_018062695.1 | XP_017918184.1 | 5 - 6 | 559 | 61 | ||
XM_018062701.1 | XP_017918190.1 | 4 - 5 | 401 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
