Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047913.1 | XP_017903402.1 | 3 - 4 | 489 | 72 | |
| XM_018047914.1 | XP_017903403.1 | 4 - 5 | 484 | 72 | ||
More Information
Set Membership: |
|

YAF2
Genomic Map