Assay Details
| Gene Symbol: LOC102170588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680604.3 | XP_005680661.1 | 3 - 4 | 1033 | 69 | |
| XM_013963982.2 | XP_013819436.1 | 4 - 5 | 1112 | 69 | ||
More Information
Set Membership: |
|

LOC102170588
Genomic Map