Assay Details
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965206.2 | XP_013820660.1 | 9 - 10 | 933 | 59 | |
XM_013965209.2 | XP_013820663.1 | 7 - 8 | 755 | 59 | ||
XM_018050985.1 | XP_017906474.1 | 9 - 10 | 1038 | 59 | ||
XM_018050986.1 | XP_017906475.1 | 10 - 11 | 1157 | 59 | ||
GenBank mRNA | JO621256.2 | - | 9 - 10 | 912 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
