Assay Details
| Gene Symbol: DAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682573.3 | XP_005682630.1 | 8 - 9 | 1320 | 59 | |
| XM_018050850.1 | XP_017906339.1 | 8 - 9 | 1080 | 59 | ||
| GenBank mRNA | JO436026.2 | - | 1 - 2 | 330 | 59 | |
More Information
Set Membership: |
|

DAPK3
Genomic Map