Assay Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050750.1 | XP_017906239.1 | 2 - 3 | 782 | 74 | |
GenBank mRNA | JO423193.2 | - | 2 - 3 | 612 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
