Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689577.3 | XP_005689634.3 | 2 - 3 | 2705 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
