Assay Details
| Gene Symbol: LOC102186042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045291.1 | XP_017900780.1 | 3 - 4 | 538 | 66 | |
| GenBank mRNA | JT678556.1 | - | 1239 | 66 | ||
More Information
Set Membership: |
|

LOC102186042
Genomic Map