Assay Details
| Gene Symbol: LIPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698184.2 | XP_005698241.2 | 4 - 5 | 535 | 74 | |
| XM_018041745.1 | XP_017897234.1 | 4 - 5 | 535 | 74 | ||
| XM_018041746.1 | XP_017897235.1 | 4 - 5 | 535 | 74 | ||
| GenBank mRNA | KA332723.1 | - | 4 - 5 | 515 | 74 | |
More Information
Set Membership: |
|

LIPK
Genomic Map