Assay Details
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057623.1 | XP_017913112.1 | 3 - 4 | 466 | 62 | |
GenBank mRNA | JO498158.2 | - | 4 - 5 | 1697 | 62 | |
KA331389.1 | - | 3 - 4 | 411 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
