Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969330.2 | XP_013824784.1 | 4 - 5 | 421 | 65 | |
XM_013969332.2 | XP_013824786.1 | 3 - 4 | 266 | 65 | ||
XM_018058207.1 | XP_017913696.1 | 4 - 5 | 435 | 65 | ||
XM_018058208.1 | XP_017913697.1 | 4 - 5 | 432 | 65 | ||
GenBank mRNA | JT697498.1 | - | 2 - 3 | 141 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
