Assay Details
Gene Symbol: LOC102184834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060680.1 | XP_017916169.1 | 19 - 20 | 2691 | 51 | |
XM_018060681.1 | XP_017916170.1 | 18 - 19 | 2634 | 51 | ||
XM_018060682.1 | XP_017916171.1 | 20 - 21 | 3995 | 51 | ||
XM_018060683.1 | XP_017916172.1 | 20 - 21 | 3995 | 51 | ||
XM_018060684.1 | XP_017916173.1 | 19 - 20 | 3938 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
