Assay Details
Gene Symbol: LOC102188791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044813.1 | XP_017900302.1 | 11 - 12 | 1198 | 88 | |
XM_018044814.1 | XP_017900303.1 | 11 - 12 | 1196 | 88 | ||
XM_018044815.1 | XP_017900304.1 | 12 - 13 | 1174 | 88 | ||
GenBank mRNA | JO610862.2 | - | 11 - 12 | 1207 | 88 | |
JO610863.2 | - | 12 - 13 | 1274 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
