Assay Details
Gene Symbol: LOC102188791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044813.1 | XP_017900302.1 | 4 - 5 | 384 | 70 | |
XM_018044814.1 | XP_017900303.1 | 4 - 5 | 382 | 70 | ||
XM_018044815.1 | XP_017900304.1 | 4 - 5 | 293 | 70 | ||
GenBank mRNA | JO610862.2 | - | 4 - 5 | 393 | 70 | |
JO610863.2 | - | 4 - 5 | 393 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018044815.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
