Assay Details
Gene Symbol: LOC102188791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044813.1 | XP_017900302.1 | 8 - 9 | 904 | 88 | |
XM_018044814.1 | XP_017900303.1 | 8 - 9 | 902 | 88 | ||
XM_018044815.1 | XP_017900304.1 | 9 - 10 | 880 | 88 | ||
GenBank mRNA | JO610862.2 | - | 8 - 9 | 913 | 88 | |
JO610863.2 | - | 9 - 10 | 980 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
