Assay Details
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064371.1 | XP_017919860.1 | 5 - 6 | 707 | 76 | |
XM_018064372.1 | XP_017919861.1 | 4 - 5 | 650 | 76 | ||
GenBank mRNA | KA341220.1 | - | 4 - 5 | 639 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
