Assay Details
Gene Symbol: LOC102168874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054837.1 | XP_017910326.1 | 2 - 3 | 280 | 63 | |
XM_018054844.1 | XP_017910333.1 | 2 - 3 | 288 | 63 | ||
GenBank mRNA | JO645987.2 | - | 2 - 3 | 470 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
