Assay Details
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975711.2 | XP_013831165.2 | 5 - 6 | 1747 | 80 | |
XM_018042493.1 | XP_017897982.1 | 5 - 6 | 1133 | 80 | ||
XM_018042494.1 | XP_017897983.1 | 5 - 6 | 1038 | 80 | ||
XM_018042495.1 | XP_017897984.1 | 5 - 6 | 1747 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
