Assay Details
| Gene Symbol: LOC102177846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059034.1 | XP_017914523.1 | 2 - 3 | 304 | 87 | |
| GenBank mRNA | JO353219.2 | - | 2 - 3 | 333 | 87 | |
More Information
Set Membership: |
|

LOC102177846
Genomic Map