Assay Details
| Gene Symbol: ORAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691414.3 | XP_005691471.2 | 1 - 2 | 746 | 75 | |
| GenBank mRNA | JO613239.2 | - | 1 - 2 | 1082 | 75 | |
| JT696857.1 | - | 2 - 3 | 881 | 75 | ||
More Information
Set Membership: |
|

ORAI1
Genomic Map