Assay Details
| Gene Symbol: LOC102182822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063286.1 | XP_017918775.1 | 3 - 4 | 251 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102182822| Gene Symbol: LOC102182822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063286.1 | XP_017918775.1 | 3 - 4 | 251 | 85 | |
Set Membership: |
|