Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697552.3 | XP_005697609.2 | 6 - 7 | 1014 | 78 | |
| XM_005697553.3 | XP_005697610.2 | 6 - 7 | 976 | 78 | ||
| XM_005697554.3 | XP_005697611.2 | 5 - 6 | 862 | 78 | ||
| XM_018041024.1 | XP_017896513.1 | 5 - 6 | 862 | 78 | ||
| GenBank mRNA | JO419569.2 | - | 2 - 3 | 513 | 78 | |
| JO438726.2 | - | 5 - 6 | 865 | 78 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map