Assay Details
| Gene Symbol: CNOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041920.1 | XP_017897409.1 | 5 - 6 | 935 | 68 | |
| XM_018041921.1 | XP_017897410.1 | 4 - 5 | 754 | 68 | ||
| GenBank mRNA | JI861764.1 | - | 3 - 4 | 454 | 68 | |
| JO427649.1 | - | 2 - 3 | 1768 | 68 | ||
| JO581271.1 | - | 1956 | 68 | |||
| JR131290.1 | - | 4587 | 68 | |||
More Information
Set Membership: |
|

CNOT7
Genomic Map