Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050823.1 | XP_017906312.1 | 2 - 3 | 631 | 63 | |
| XM_018050824.1 | XP_017906313.1 | 2 - 3 | 634 | 63 | ||
| GenBank mRNA | JO434949.2 | - | 3 - 4 | 1075 | 63 | |
More Information
Set Membership: |
|

DOHH
Genomic Map