Assay Details
Gene Symbol: GTF3C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966432.2 | XP_013821886.2 | 5 - 6 | 430 | 79 | |
GenBank mRNA | JO644545.2 | - | 277 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
