Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057626.1 | XP_017913115.1 | 2 - 3 | 294 | 60 | |
XM_018057627.1 | XP_017913116.1 | 2 - 3 | 334 | 60 | ||
GenBank mRNA | JO639050.2 | - | 2 - 3 | 241 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
