Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058230.1 | XP_017913719.1 | 3 - 4 | 600 | 61 | |
XM_018058231.1 | XP_017913720.1 | 3 - 4 | 525 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
