Assay Details
| Gene Symbol: MEAK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061792.1 | XP_017917281.1 | 2 - 3 | 531 | 63 | |
| XM_018061793.1 | XP_017917282.1 | 3 - 4 | 507 | 63 | ||
| XM_018061794.1 | XP_017917283.1 | 3 - 4 | 537 | 63 | ||
More Information
Set Membership: |
|

MEAK7
Genomic Map