Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061792.1 | XP_017917281.1 | 6 - 7 | 1412 | 100 | |
XM_018061793.1 | XP_017917282.1 | 7 - 8 | 1388 | 100 | ||
XM_018061794.1 | XP_017917283.1 | 7 - 8 | 1418 | 100 | ||
GenBank mRNA | JO416227.2 | - | 3 - 4 | 509 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
