Assay Details
Gene Symbol: LOC108633813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039802.1 | XP_017895291.1 | 2 - 3 | 262 | 59 | |
GenBank mRNA | JO627466.2 | - | 1 - 2 | 75 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
