Assay Details
Gene Symbol: LOC108633813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039802.1 | XP_017895291.1 | 3 - 4 | 471 | 91 | |
GenBank mRNA | JO627466.2 | - | 2 - 3 | 284 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
