Assay Details
Gene Symbol: LOC108633813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039802.1 | XP_017895291.1 | 4 - 5 | 611 | 69 | |
GenBank mRNA | JO627466.2 | - | 3 - 4 | 424 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
