Assay Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674962.3 | XP_005675019.1 | 1 - 2 | 909 | 95 | |
| XM_005674963.3 | XP_005675020.1 | 1 - 2 | 909 | 95 | ||
| XM_005674964.3 | XP_005675021.1 | 1 - 2 | 909 | 95 | ||
| GenBank mRNA | JO358421.2 | - | 1 - 2 | 811 | 95 | |
| JO374747.2 | - | 1 - 2 | 382 | 95 | ||
| JO374748.2 | - | 1 - 2 | 382 | 95 | ||
| JO553461.2 | - | 1 - 2 | 148 | 95 | ||
More Information
Set Membership: |
|

POPDC2
Genomic Map