Assay Details
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675616.3 | XP_005675673.2 | 4 - 5 | 2223 | 87 | |
XM_005675617.3 | XP_005675674.2 | 4 - 5 | 1820 | 87 | ||
XM_005675618.3 | XP_005675675.2 | 3 - 4 | 2052 | 87 | ||
XM_005675619.3 | XP_005675676.2 | 4 - 5 | 2224 | 87 | ||
XM_013966561.2 | XP_013822015.2 | 4 - 5 | 1861 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
