Assay Details
Gene Symbol: SARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677960.3 | XP_005678017.1 | 7 - 8 | 1088 | 64 | |
GenBank mRNA | JO503274.2 | - | 4 - 5 | 869 | 64 | |
JO556578.2 | - | 4 - 5 | 832 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
