Assay Details
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680682.3 | XP_005680739.1 | 2 - 3 | 1035 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
