Assay Details
Gene Symbol: MFAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680893.3 | XP_005680950.1 | 7 - 8 | 488 | 65 | |
GenBank mRNA | JT762682.1 | - | 7 - 8 | 609 | 65 | |
KA328244.1 | - | 7 - 8 | 410 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
