Assay Details
| Gene Symbol: LOC102174373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681282.3 | XP_005681339.2 | 3 - 4 | 1749 | 71 | |
| XM_018049933.1 | XP_017905422.1 | 2 - 3 | 1069 | 71 | ||
More Information
Set Membership: |
|

LOC102174373
Genomic Map