Assay Details
Gene Symbol: LOC102171051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051429.1 | XP_017906918.1 | 2 - 3 | 661 | 78 | |
GenBank mRNA | JO521050.2 | - | 2 - 3 | 319 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
