Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050713.1 | XP_017906202.1 | 4 - 5 | 744 | 95 | |
XM_018050714.1 | XP_017906203.1 | 5 - 6 | 845 | 95 | ||
XM_018050715.1 | XP_017906204.1 | 6 - 7 | 963 | 95 | ||
XM_018050716.1 | XP_017906205.1 | 4 - 5 | 746 | 95 | ||
XM_018050717.1 | XP_017906206.1 | 4 - 5 | 621 | 95 | ||
XM_018050718.1 | XP_017906207.1 | 4 - 5 | 682 | 95 | ||
XM_018050719.1 | XP_017906208.1 | 3 - 4 | 581 | 95 | ||
GenBank mRNA | JO425921.1 | - | 2 - 3 | 2633 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
