Assay Details
| Gene Symbol: FGF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683284.3 | XP_005683341.2 | 2 - 3 | 414 | 61 | |
| GenBank mRNA | JO624544.2 | - | 2 - 3 | 303 | 61 | |
| KA331441.1 | - | 2 - 3 | 396 | 61 | ||
More Information
Set Membership: |
|

FGF22
Genomic Map