Assay Details
| Gene Symbol: PPP1R14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054436.1 | XP_017909925.1 | 3 - 4 | 471 | 73 | |
| XM_018054437.1 | XP_017909926.1 | 2 - 3 | 328 | 73 | ||
More Information
Set Membership: |
|

PPP1R14D
Genomic Map