Assay Details
Gene Symbol: SRMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057532.1 | XP_017913021.1 | 1 - 2 | 600 | 62 | |
GenBank mRNA | KA339345.1 | - | 7 - 8 | 3143 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
