Assay Details
| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042313.1 | XP_017897802.1 | 4 - 5 | 2168 | 113 | |
| XM_018042314.1 | XP_017897803.1 | 5 - 6 | 2061 | 113 | ||
| XM_018042315.1 | XP_017897804.1 | 6 - 7 | 1305 | 113 | ||
| XM_018042316.1 | XP_017897805.1 | 5 - 6 | 2123 | 113 | ||
| XM_018042317.1 | XP_017897806.1 | 5 - 6 | 1233 | 113 | ||
| XM_018042319.1 | XP_017897808.1 | 5 - 6 | 1021 | 113 | ||
| XM_018042320.1 | XP_017897809.1 | 5 - 6 | 511 | 113 | ||
| XM_018042321.1 | XP_017897810.1 | 4 - 5 | 2193 | 113 | ||
More Information
Set Membership: |
|

ZNF25
Genomic Map