Assay Details
| Gene Symbol: NUDT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966112.2 | XP_013821566.1 | 2 - 3 | 907 | 63 | |
| GenBank mRNA | JO647698.2 | - | 1 - 2 | 1119 | 63 | |
More Information
Set Membership: |
|

NUDT18
Genomic Map