Assay Details
Gene Symbol: TENT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052911.1 | XP_017908400.1 | 2 - 3 | 850 | 100 | |
XM_018052912.1 | XP_017908401.1 | 2 - 3 | 794 | 100 | ||
XM_018052913.1 | XP_017908402.1 | 2 - 3 | 858 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
