Assay Details
| Gene Symbol: LIPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969771.2 | XP_013825225.2 | 1 - 2 | 463 | 56 | |
| GenBank mRNA | JT708419.1 | - | 3 - 4 | 690 | 56 | |
More Information
Set Membership: |
|

LIPT2
Genomic Map